Assay Design



Gene Transcripts

Gene Symbol
KCNK10
Entrez Gene ID
100009503
Gene Name
potassium two pore domain channel subfamily K member 10
Gene Aliases
-
Chromosome Location
Chr.20: 17057670 - 17196865 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082736.1 NP_001076205.1 5 - 6 907 100
GenBank mRNA AY553324.1 - 5 - 6 907 100

View Full Product Details