Assay Design



Gene Transcripts

Gene Symbol
LY96
Entrez Gene ID
100009589
Gene Name
lymphocyte antigen 96
Gene Aliases
-
Chromosome Location
Chr.3: 89728214 - 89773408 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082787.2 NP_001076256.1 4 - 5 449 94
XM_008255647.2 XP_008253869.1 5 - 6 644 94
GenBank mRNA AY101395.1 - 4 - 5 451 94

View Full Product Details