Assay Design



Gene Transcripts

Gene Symbol
TF
Entrez Gene ID
100009267
Gene Name
transferrin
Gene Aliases
TRF
Chromosome Location
Chr.14: 27729544 - 27758319 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101694.1 NP_001095164.1 11 - 12 1347 63
GenBank mRNA X58533.1 - - 1347 63

View Full Product Details