Assay Design



Gene Transcripts

Gene Symbol
ORM1
Entrez Gene ID
100009268
Gene Name
orosomucoid 1
Gene Aliases
-
Chromosome Location
Chr.Unknown: 13299650 - 13303016 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101695.1 NP_001095165.1 - 364 68
GenBank mRNA X58727.1 - - 364 68

View Full Product Details