Assay Design



Gene Transcripts

Gene Symbol
KCNC1
Entrez Gene ID
100008968
Gene Name
potassium voltage-gated channel subfamily C member 1
Gene Aliases
Kv3.1
Chromosome Location
Chr.Unknown: 1754378 - 1791145 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082182.1 NP_001075651.1 - 1448 67
XM_017350898.1 XP_017206387.1 - 2010 67
GenBank mRNA AY928179.1 - - 1448 67

View Full Product Details