Assay Design



Gene Transcripts

Gene Symbol
LHB
Entrez Gene ID
100009427
Gene Name
luteinizing hormone subunit beta
Gene Aliases
-
Chromosome Location
Chr.Unknown: 51898 - 52790 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082695.1 NP_001076164.1 - 53 90
GenBank mRNA AB235913.1 - - 53 90
AB235914.1 - - 53 90
AY614703.2 - - 53 90

View Full Product Details