Assay Design



Gene Transcripts

Gene Symbol
GPX1
Entrez Gene ID
100009258
Gene Name
glutathione peroxidase 1
Gene Aliases
-
Chromosome Location
Chr.9: 16889539 - 16890546 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085444.1 NP_001078913.1 1 - 2 260 75
GenBank mRNA X13837.1 - - 260 75

View Full Product Details