Assay Design



Gene Transcripts

Gene Symbol
MT3
Entrez Gene ID
100009025
Gene Name
metallothionein 3
Gene Aliases
GIF, MT-III
Chromosome Location
Chr.5: 12893395 - 12894753 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082220.2 NP_001075689.2 2 - 3 140 77
GenBank mRNA DQ304562.1 - 2 - 3 93 77

View Full Product Details