Assay Design



Gene Transcripts

Gene Symbol
CRP
Entrez Gene ID
100009091
Gene Name
C-reactive protein
Gene Aliases
PTX1
Chromosome Location
Chr.13: 33405195 - 33407744 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082265.1 NP_001075734.1 2 - 2 1949 98
GenBank mRNA M13497.1 - 2 - 2 1949 98

View Full Product Details