Assay Design



Gene Transcripts

Gene Symbol
CD14
Entrez Gene ID
100008983
Gene Name
CD14 molecule
Gene Aliases
-
Chromosome Location
Chr.3: 22754430 - 22755771 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082195.2 NP_001075664.1 2 - 2 1154 95
GenBank mRNA D16545.1 - 2 - 2 1155 95

View Full Product Details