Assay Design



Gene Transcripts

Gene Symbol
LTA
Entrez Gene ID
100328931
Gene Name
lymphotoxin alpha
Gene Aliases
TNF-beta, TNLG1E
Chromosome Location
Chr.12: 20396271 - 20398217 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171119.1 NP_001164590.1 2 - 3 234 101

View Full Product Details