Assay Design



Gene Transcripts

Gene Symbol
CYP3A6
Entrez Gene ID
100328954
Gene Name
cytochrome P450 3A6
Gene Aliases
-
Chromosome Location
Chr.Unknown: 607005 - 640099 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171268.2 NP_001164739.1 - 210 100
GenBank mRNA J05034.1 - - 211 100

View Full Product Details