Assay Design



Gene Transcripts

Gene Symbol
SERPINB12
Entrez Gene ID
100328643
Gene Name
serpin family B member 12
Gene Aliases
-
Chromosome Location
Chr.9: 103780438 - 103806410 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171282.1 NP_001164753.1 2 - 3 241 77

View Full Product Details