Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171282.1 | NP_001164753.1 | 6 - 7 | 811 | 80 |
XM_008261379.2 | XP_008259601.1 | 7 - 8 | 954 | 80 | |
XM_008261380.2 | XP_008259602.1 | 7 - 8 | 994 | 80 | |
XM_017343648.1 | XP_017199137.1 | 8 - 9 | 1054 | 80 |