Assay Design



Gene Transcripts

Gene Symbol
MAP1A
Entrez Gene ID
100328681
Gene Name
microtubule associated protein 1A
Gene Aliases
-
Chromosome Location
Chr.17: 28723892 - 28744156 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171311.1 NP_001164782.1 3 - 4 8229 95
XM_008268819.2 XP_008267041.1 6 - 7 9123 95

View Full Product Details