Assay Design



Gene Transcripts

Gene Symbol
SLC5A4
Entrez Gene ID
100328735
Gene Name
solute carrier family 5 member 4
Gene Aliases
-
Chromosome Location
Chr.21: 4484605 - 4526392 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171357.1 NP_001164828.1 8 - 9 886 100

View Full Product Details