Assay Design



Gene Transcripts

Gene Symbol
BPIFC
Entrez Gene ID
100328738
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
Chromosome Location
Chr.4: 88630325 - 88697818 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171359.1 NP_001164830.1 4 - 5 531 65
XM_008257060.2 XP_008255282.1 5 - 6 639 65
XM_008257061.2 XP_008255283.1 6 - 7 1985 65
XM_008257062.2 XP_008255284.1 4 - 5 511 65

View Full Product Details