Assay Design



Gene Transcripts

Gene Symbol
BPIFC
Entrez Gene ID
100328738
Gene Name
BPI fold containing family C
Gene Aliases
BPIL2
Chromosome Location
Chr.4: 88630325 - 88697818 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171359.1 NP_001164830.1 9 - 10 977 96
XM_008257060.2 XP_008255282.1 10 - 11 1085 96
XM_008257061.2 XP_008255283.1 11 - 12 2431 96
XM_008257062.2 XP_008255284.1 9 - 10 957 96

View Full Product Details