Assay Design



Gene Transcripts

Gene Symbol
MYADM
Entrez Gene ID
100328773
Gene Name
myeloid associated differentiation marker
Gene Aliases
-
Chromosome Location
Chr.Unknown: 159181 - 165761 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171388.1 NP_001164859.1 - 853 80
XM_008251642.2 XP_008249864.1 - 1041 80

View Full Product Details