Assay Design



Gene Transcripts

Gene Symbol
OR51I2
Entrez Gene ID
100328868
Gene Name
olfactory receptor, family 51, subfamily I, member 2
Gene Aliases
-
Chromosome Location
Chr.1: 146405903 - 146409129 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171461.1 NP_001164932.1 1 - 1 741 82
XM_017343433.1 XP_017198922.1 2 - 2 759 82

View Full Product Details