Assay Design



Gene Transcripts

Gene Symbol
UBQLN3
Entrez Gene ID
100328875
Gene Name
ubiquilin 3
Gene Aliases
-
Chromosome Location
Chr.1: 146527227 - 146529933 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171468.1 NP_001164939.1 1 - 1 1846 77
XM_008258925.2 XP_008257147.1 2 - 2 2048 77

View Full Product Details