Assay Design



Gene Transcripts

Gene Symbol
CFAP298
Entrez Gene ID
100328921
Gene Name
cilia and flagella associated protein 298
Gene Aliases
C14H21orf59, C21orf59, RA_m005_jsm24C07r
Chromosome Location
Chr.14: 162600908 - 162609228 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171500.1 NP_001164971.1 1 - 2 136 62

View Full Product Details