Assay Design



Gene Transcripts

Gene Symbol
GTF2A1L
Entrez Gene ID
100534011
Gene Name
general transcription factor IIA subunit 1 like
Gene Aliases
-
Chromosome Location
Chr.2: 137700783 - 137771420 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204847.1 NP_001191776.1 6 - 7 1047 111

View Full Product Details