Gene Transcripts

Gene Symbol
Scaper
Entrez Gene ID
117521
Gene Name
S-phase cyclin A-associated protein in the ER
Gene Aliases
Zfp291, Znf291
Chromosome Location
Chr. 8: 60127039 - 60593566 on Build Rnor_6.0
UniGene ID
Rn.232456
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766363.1 XP_008764585.1 19, 20 2491
XM_008766364.1 XP_008764586.1 19, 20 2596
XM_008766365.1 XP_008764587.1 19, 20 2473

View Full Product Details