Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012993.2 | NP_037125.1 | 26 | 2942 | |
XM_006238499.2 | XP_006238561.1 | 28 | 3280 | ||
XM_006238500.2 | XP_006238562.1 | 27 | 3144 | ||
GenBank mRNA | FQ218095.1 | - | 26 | 3065 | |
L27124.1 | - | 26 | 2943 | ||
X93208.1 | - | 28 | 3147 |