Gene Transcripts

Gene Symbol
Chst15
Entrez Gene ID
286974
Gene Name
carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15
Gene Aliases
GalNAc4S6ST, Galnac4s-6st
Chromosome Location
Chr. 1: 204243396 - 204328970 on Build Rnor_6.0
UniGene ID
Rn.217784
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173310.3 NP_775432.3 4, 5 1207
XM_006230430.2 XP_006230492.1 5, 6 2774
XM_006230431.2 XP_006230493.1 5, 6 3514
XM_006230433.2 XP_006230495.1 5, 6 3009
XM_008759868.1 XP_008758090.1 5, 6 1770
GenBank mRNA AB086953.1 - 4, 5 1207

View Full Product Details