Gene Transcripts

Gene Symbol
Fbxw9
Entrez Gene ID
288921
Gene Name
F-box and WD repeat domain containing 9
Gene Aliases
Chromosome Location
Chr. 19: 26173771 - 26180675 on Build Rnor_6.0
UniGene ID
Rn.29201
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081634.1 NP_001075103.1 8 1247
GenBank mRNA BC166590.1 - 5 1447

View Full Product Details