Gene Transcripts

Gene Symbol
Stab1
Entrez Gene ID
100363145
Gene Name
stabilin 1
Gene Aliases
Chromosome Location
Chr. 16: 7220434 - 7250935 on Build Rnor_6.0
UniGene ID
Rn.144136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771053.1 XP_008769275.1 64 7554
XM_008771054.1 XP_008769276.1 64 7551
XM_008771055.1 XP_008769277.1 65 7386

View Full Product Details