Gene Transcripts

Gene Symbol
Chat
Entrez Gene ID
290567
Gene Name
choline O-acetyltransferase
Gene Aliases
Chromosome Location
Chr. 16: 8576858 - 8583314 on Build Rnor_6.0
UniGene ID
Rn.45116
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170593.1 NP_001164064.1 1375

View Full Product Details