Gene Transcripts

Gene Symbol
Trim27
Entrez Gene ID
291171
Gene Name
tripartite motif-containing 27
Gene Aliases
Rfp1
Chromosome Location
Chr. 17: 45613863 - 45626212 on Build Rnor_6.0
UniGene ID
Rn.8393
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134974.1 NP_001128446.1 3 916
GenBank mRNA BC169072.1 - 3 916

View Full Product Details