Gene Transcripts

Gene Symbol
Ugt3a2
Entrez Gene ID
294793
Gene Name
UDP glycosyltransferase 3 family, polypeptide A2
Gene Aliases
RGD1564365
Chromosome Location
Chr. 2: 58666337 - 58682212 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008760833.1 XP_008759055.1 4 955

View Full Product Details