Gene Transcripts

Gene Symbol
Fam73b
Entrez Gene ID
296623
Gene Name
family with sequence similarity 73, member B
Gene Aliases
RGD1310887
Chromosome Location
Chr. 3: 8928495 - 8951189 on Build Rnor_6.0
UniGene ID
Rn.17259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106566.1 NP_001100036.1 1, 2 55
XM_006233781.1 XP_006233843.1 1, 2 104
GenBank mRNA FQ212119.1 - 1, 2 23
FQ231217.1 - 1, 2 59

View Full Product Details