Gene Transcripts

Gene Symbol
Txnrd3
Entrez Gene ID
297437
Gene Name
thioredoxin reductase 3
Gene Aliases
Chromosome Location
Chr. 4: 121612332 - 121650543 on Build Rnor_6.0
UniGene ID
Rn.48039
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106609.2 NP_001100079.2 10 1405
NM_001184712.1 NP_001171641.1 10 1405
GenBank mRNA CB585038.1 - 3 390
CK603679.1 - 2 101

View Full Product Details