Gene Transcripts

Gene Symbol
Tfap2d
Entrez Gene ID
301284
Gene Name
transcription factor AP-2 delta
Gene Aliases
Tcfap2d
Chromosome Location
Chr. 9: 25304876 - 25365151 on Build Rnor_6.0
UniGene ID
Rn.163046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106895.1 NP_001100365.1 6 794

View Full Product Details