Gene Transcripts

Gene Symbol
Unc45b
Entrez Gene ID
303373
Gene Name
unc-45 homolog B (C. elegans)
Gene Aliases
Cmya4
Chromosome Location
Chr. 10: 70262342 - 70290445 on Build Rnor_6.0
UniGene ID
Rn.61093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107028.1 NP_001100498.1 6 657
XM_006246996.2 XP_006247058.1 6 654

View Full Product Details