Gene Transcripts

Gene Symbol
Ube2o
Entrez Gene ID
303689
Gene Name
ubiquitin-conjugating enzyme E2O
Gene Aliases
RGD1310297
Chromosome Location
Chr. 10: 105505721 - 105553137 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_221132.8 XP_221132.5 15 2930

View Full Product Details