Gene Transcripts

Gene Symbol
Trim5
Entrez Gene ID
308906
Gene Name
tripartite motif-containing 5
Gene Aliases
RGD1304579
Chromosome Location
Chr. 1: 169446774 - 169463760 on Build Rnor_6.0
UniGene ID
Rn.12117
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014023.1 NP_001014045.1 2 489
XM_006229912.2 XP_006229974.1 2 975
GenBank mRNA BC078926.1 - 2 489

View Full Product Details