Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079706.1 | NP_001073174.1 | 5, 6 | 545 | |
XM_006230111.2 | XP_006230173.1 | 4, 5 | 475 | ||
XM_006230121.2 | XP_006230183.1 | 1, 2 | 427 | ||
XM_008759751.1 | XP_008757973.1 | 4, 5 | 481 | ||
XM_008759752.1 | XP_008757974.1 | 4, 5 | 766 | ||
XM_008759753.1 | XP_008757975.1 | 4, 5 | 767 | ||
XM_008759754.1 | XP_008757976.1 | 4, 5 | 481 | ||
XM_008759755.1 | XP_008757977.1 | 4, 5 | 766 | ||
XM_008759756.1 | XP_008757978.1 | 4, 5 | 766 | ||
XM_008759757.1 | XP_008757979.1 | 2, 3 | 526 | ||
XM_008759758.1 | XP_008757980.1 | 4, 5 | 768 | ||
XM_008759759.1 | XP_008757981.1 | 3, 4 | 254 | ||
XM_008759760.1 | XP_008757982.1 | 4, 5 | 481 | ||
GenBank mRNA | BC128775.1 | - | 5, 6 | 545 |