Gene Transcripts

Gene Symbol
LOC102556290
Entrez Gene ID
102556290
Gene Name
nipped-B-like protein B-like
Gene Aliases
Chromosome Location
Chr. 1: 220891052 - 220913279 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006230945.2 XP_006231007.1 1 773

View Full Product Details