Gene Transcripts

Gene Symbol
Nmi
Entrez Gene ID
311021
Gene Name
N-myc (and STAT) interactor
Gene Aliases
Chromosome Location
Chr. 3: 37458891 - 37480984 on Build Rnor_6.0
UniGene ID
Rn.18496
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034148.1 NP_001029320.1 686
GenBank mRNA BC100089.1 - 686

View Full Product Details