Gene Transcripts

Gene Symbol
Wdtc1
Entrez Gene ID
313020
Gene Name
WD and tetratricopeptide repeats 1
Gene Aliases
Chromosome Location
Chr. 5: 151487572 - 151536812 on Build Rnor_6.0
UniGene ID
Rn.16646
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107908.1 NP_001101378.1 7 1006
XM_008764134.1 XP_008762356.1 7 1145
XM_008764135.1 XP_008762357.1 7 802

View Full Product Details