Gene Transcripts

Gene Symbol
Zmynd12
Entrez Gene ID
313552
Gene Name
zinc finger, MYND-type containing 12
Gene Aliases
Chromosome Location
Chr. 5: 138470173 - 138505928 on Build Rnor_6.0
UniGene ID
Rn.60862
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107970.1 NP_001101440.1 5, 6 811
XM_006238759.2 XP_006238821.1 5, 6 826
XM_006238760.2 XP_006238822.1 5, 6 824

View Full Product Details