Gene Transcripts

Gene Symbol
Ptchd2
Entrez Gene ID
313705
Gene Name
patched domain containing 2
Gene Aliases
RGD1305820
Chromosome Location
Chr. 5: 165051522 - 165099823 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750087.3 XP_003750135.1 7 2041

View Full Product Details