Gene Transcripts

Gene Symbol
RGD1562284
Entrez Gene ID
313837
Gene Name
similar to Glutaminyl-peptide cyclotransferase precursor (QC)
Gene Aliases
Chromosome Location
Chr. 6: 1657114 - 1689575 on Build Rnor_6.0
UniGene ID
Rn.7474
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006239632.2 XP_006239694.1 2 441
GenBank mRNA FQ229106.1 - 2 235

View Full Product Details