Gene Transcripts

Gene Symbol
Plekhh2
Entrez Gene ID
313866
Gene Name
pleckstrin homology domain containing, family H (with MyTH4 domain) member 2
Gene Aliases
RGD1304935
Chromosome Location
Chr. 6: 7784737 - 7895986 on Build Rnor_6.0
UniGene ID
Rn.204776
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191770.1 NP_001178699.1 28 4247
XM_008764459.1 XP_008762681.1 29 4676
XM_008764460.1 XP_008762682.1 29 4384
XM_008764461.1 XP_008762683.1 29 4376
XM_008764462.1 XP_008762684.1 28 4755
XM_008764463.1 XP_008762685.1 27 4181

View Full Product Details