Gene Transcripts

Gene Symbol
Wdr75
Entrez Gene ID
314545
Gene Name
WD repeat domain 75
Gene Aliases
LRRG00118
Chromosome Location
Chr. 9: 52687850 - 52717808 on Build Rnor_6.0
UniGene ID
Rn.38742
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047889.1 NP_001041354.1 1 77
XM_006244892.2 XP_006244954.1 4 350
GenBank mRNA AY539869.1 - 1 77

View Full Product Details