Gene Transcripts

Gene Symbol
Glyr1
Entrez Gene ID
360477
Gene Name
glyoxylate reductase 1 homolog (Arabidopsis)
Gene Aliases
3930401k13rik, N-pac, RGD1309459
Chromosome Location
Chr. 10: 10725761 - 10761282 on Build Rnor_6.0
UniGene ID
Rn.2639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007800.1 NP_001007801.1 5 358
XM_006245803.2 XP_006245865.1 5 427
GenBank mRNA BC085931.1 - 5 369

View Full Product Details