Gene Transcripts

Gene Symbol
Tbc1d1
Entrez Gene ID
360937
Gene Name
TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1
Gene Aliases
Chromosome Location
Chr. 14: 45598753 - 45797649 on Build Rnor_6.0
UniGene ID
Rn.234365
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251004.2 XP_006251066.1 8 1623
XM_006251005.2 XP_006251067.1 8 1623
XM_006251006.2 XP_006251068.1 7 1017
XM_006251007.1 XP_006251069.1 7 1041
XM_341215.7 XP_341216.4 8 1623

View Full Product Details