Gene Transcripts

Gene Symbol
Rnaseh2b
Entrez Gene ID
361056
Gene Name
ribonuclease H2, subunit B
Gene Aliases
Chromosome Location
Chr. 15: 45422010 - 45472792 on Build Rnor_6.0
UniGene ID
Rn.16136
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007007.1 NP_001007008.1 5 385
XM_008770762.1 XP_008768984.1 5 2458
XM_008770763.1 XP_008768985.1 5 2458
XM_008770764.1 XP_008768986.1 5 2460
XM_008770765.1 XP_008768987.1 5 2458
GenBank mRNA BC083791.1 - 5 385
FQ220108.1 - 5 412

View Full Product Details