Gene Transcripts

Gene Symbol
Snx32
Entrez Gene ID
361708
Gene Name
sorting nexin 32
Gene Aliases
RGD1560591, Snx6b
Chromosome Location
Chr. 1: 220873096 - 220890815 on Build Rnor_6.0
UniGene ID
Rn.40837
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006230940.2 XP_006231002.1 12 1453
XM_006230941.2 XP_006231003.1 12 1432
XM_008760201.1 XP_008758423.1 10 1958
XM_008760202.1 XP_008758424.1 11 1853
XM_341993.7 XP_341994.5 13 1377

View Full Product Details